ASTM D6584: GC Analysis of Glycerin Impurity in B100 Biodiesel on MET-Biodiesel
ASTM D6584: GC Analysis of Glycerin Impurity in B100 Biodiesel on MET-Biodiesel
Conditions
column MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm ID guard (28668-U)
oven 50 °C (1 min), 15 °C/min to 180 °C, 7 °C/min to 230 °C, 30 °C/min to 380 °C (10 min)
carrier gas helium, 3.0 mL/min
sample B100 Biodiesel plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA then diluted in heptane
injection 1 µL, cold on-column
detector FID, 380 °C
Description
Categories Analytical Chromatography, GC Applications, Glycerides
Featured Industry Biofuels
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Butanetriol
  • Diolein
  • Glycerin
  • Monoolein
  • Tricaprin
  • Triolein