GC Analysis of 1-Aminoindan Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-DP
GC Analysis of 1-Aminoindan Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-DP
Conditions
column Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 µm (78033AST)
oven 160 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: 1-aminoindan enantiomers (N-TFA derivatives)
inj. temp. 250 °C
Description
Categories Analytical Chromatography, GC Applications, Chiral Applications, Pharmaceutical (small molecule), Clinical, Forensic & Toxicology, Antiparkinson Drugs, Enantiomers, Therapeutic Drug Monitoring
Featured Industry Clinical
Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes