GC Analysis of 1-Phenylethylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX® B-PM
GC Analysis of 1-Phenylethylamine Enantiomers (N-Acetyl Derivatives) on Astec® CHIRALDEX® B-PM
Conditions
column Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 µm (76023AST)
oven 130 °C
carrier gas helium, 30 psi
sample 3 mg/mL in methylene chloridepeak 1: S(-)-1-phenylethylamine (N-TFA derivative)peak 2: R(+)-1-phenylethylamine (N-TFA derivative)
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Can reverse elution order by using the N-TFA derivative on CHIRALDEX G-PN or B-DM columns.
Categories Analytical Chromatography, GC Applications, Anilines, Amines, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes