GC Analysis of 17 Dioxins + 7 IS/SS on Equity®-5, Fast GC Analysis
GC Analysis of 17 Dioxins + 7 IS/SS on Equity®-5, Fast GC Analysis
Conditions
column Equity-5, 15 m x 0.10 mm I.D., 0.10 µm (28083-U)
oven 150 °C (1 min), 45 °C/min to 300 °C (4 min)
carrier gas hydrogen, 50 cm/sec constant
sample Dioxin standard, each analyte at 500-2500 ppb in nonane
injection 0.5 µL, splitless (1.0 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector ECD, 325 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Agriculture
Environmental
Food and Beverages
Industrial Hygiene
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes