GC Analysis of 2-Aminobutane (2-Butylamine) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-TA
GC Analysis of 2-Aminobutane (2-Butylamine) Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 80 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: 2-aminobutane enantiomers (N-TFA derivatives)
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Amines, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • sec-Butylamine