GC Analysis of a 17-Component Dioxin Congener Mix on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of a 17-Component Dioxin Congener Mix on SLB®-5ms (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 15 m x 0.10 mm I.D., 0.10 µm (28466-U)
oven 150 °C (1 min), 35 °C/min to 340 °C (1 min)
carrier gas hydrogen, 45 cm/sec, constant
sample 17-component 2,3,7,8-substituted dioxin standard, 100-500 ppb in n-nonane
injection 1 µLsplitless (1 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector ECD, 340 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Agriculture
Environmental
Food and Beverages
Industrial Hygiene
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes