GC Analysis of a 17-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of a 17-Component Pesticide Mix on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 °C (1 min), 80 °C/min to 160 °C, 5 °C/min to 340 °C (2 min)
carrier gas helium, 0.52 mL/min
sample 17-component pesticide mix, each analyte at 0.8-2.0 µg/mL in hexane:acetone (50:50)
injection 0.5 µL10:1 split
inj. temp. 280 °C
MSD interface 280 °C
scan range 45-470 m/z
Description
Categories Analytical Chromatography, GC Applications, Pesticides
Featured Industry Agriculture
Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes