GC Analysis of a 37-Component FAME Mix on Omegawax®
GC Analysis of a 37-Component FAME Mix on Omegawax®
Conditions
column Omegawax 250, 30 m x 0.25 mm I.D., 0.25 µm (24136)
oven 50 °C, 3.0 °C/min to 280 °C
carrier gas helium, 40 cm/sec constant
sample Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
injection 1 µL50:1 split
inj. temp. 240 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information Omegawax is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes