GC Analysis of a 37-Component FAME Mix on SLB®-IL100
GC Analysis of a 37-Component FAME Mix on SLB®-IL100
Conditions
column SLB-IL100, 30 m x 0.25 mm I.D., 0.20 µm (28884-U)
oven 50 °C, 3.0 °C/min to 240 °C
carrier gas helium, 40 cm/sec constant
sample Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
injection 1 µL50:1 split
inj. temp. 240 °C
detector FID, 240 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes