GC Analysis of a 38-Component Fatty Acid Methyl Ester (FAME) Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of a 38-Component Fatty Acid Methyl Ester (FAME) Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
column PEG 3, 30 m x 0.25 mm I.D., 0.25 µm
oven 170 °C, 1 °C/min to 225 °C
carrier gas helium, 1.2 mL/min
sample Supelco 37-Component FAME Mix (47885-U) + C22:5n3, in methylene chloride
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner design
liner 1.0 µL100:1 split
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical