GC Analysis of a 6-Component Organophosphorus Pesticide (OP-Pest) Mix on SPB®-608
GC Analysis of a 6-Component Organophosphorus Pesticide (OP-Pest) Mix on SPB®-608
Conditions
column SPB®-608, 30 m x 0.53 mm I.D., 0.50 µm (25312)
oven 125 °C (0.5 min), 6 C/min to 230 °C (2 min), 3 °C/min to 245 °C
carrier gas helium, 8 mL/min
sample each analyte at 1 µg/mL
injection direct
detector FPD, 280 °C
Description
Categories Analytical Chromatography, Food & Beverage Analysis, GC Applications, Pesticide and Metabolite Residues, Applications (Chromatograms), Pesticides
Featured Industry Agriculture
Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Bromophos
  • Ethion
  • Ethyl parathion
  • Etrimfos
  • Methacrifos
  • Methyl parathion