GC Analysis of a 6-Component Tetrachlorodibenzo-p-dioxin (TCDD) Mix on SLB®-5ms
GC Analysis of a 6-Component Tetrachlorodibenzo-p-dioxin (TCDD) Mix on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 170 °C (1 min), 8 °C/min to 270 °C (10 min)
carrier gas helium, 37 cm/sec., constant
sample TCDD standard, 1500 ppb in n-dodecane
injection 1 µL, splitless (1 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
MSD interface 270 °C
scan range SIM, m/z = 320, 322, 324
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Agriculture
Environmental
Food and Beverages
Industrial Hygiene
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3,4-Tetrachlorodibenzo-p-dioxin
  • 1,2,3,7-Tetrachlorodibenzo-p-dioxin
  • 1,2,3,8-Tetrachlorodibenzo-p-dioxin
  • 1,2,7,8-Tetrachlorodibenzo-p-dioxin
  • 1,4,7,8-Tetrachlorodibenzo-p-dioxin
  • 2,3,7,8-Tetrachlorodibenzo-p-dioxin