GC Analysis of a QC Test Mix on SLB®-IL60i
GC Analysis of a QC Test Mix on SLB®-IL60i
Conditions
column SLB-IL60i, 30 m x 0.25 mm I.D., 0.20 µm (29832-U)
oven 130 °C
carrier gas hydrogen, 40 cm/sec
sample polar column text mix (47302), 9 analytes, each at 500 µg/mL in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 280 °C
Description
Analysis Note The i-series of ionic liquid capillary GC columns are characterized by their improved inertness for polar analytes. This application shows that the SLB IL60i can generate great peak shapes for analytes with various functionality.
Featured Industry General Analytical