GC Analysis of Alcohols on PAG
GC Analysis of Alcohols on PAG
Conditions
column PAG, 30 m x 0.25 mm I.D., 0.25 µm (24223)
oven 60 °C
carrier gas helium, 20 cm/sec
sample 0.5 - 17 mM alcohols in water
injection 1 µL
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Alcohols
Featured Industry Chemicals and Industrial Polymers
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Butanol
  • 1-Pentanol
  • 1-Propanol
  • 2-Butanol
  • 2-Methyl-1-butanol
  • 2-Methyl-1-propanol
  • 2-Methyl-2-butanol
  • 2-Methyl-2-propanol
  • 2-Pentanol
  • 3-Methyl-1-butanol
  • 3-Pentanol
  • Ethanol
  • Isopropanol
  • Methanol