GC Analysis of Amino Acids (as TBDMS Derivatives) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
GC Analysis of Amino Acids (as TBDMS Derivatives) on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Conditions
column SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
oven 100 °C (1 min), 35 °C/min to 290 °C (3 min), 40 °C/min to 360 °C
carrier gas helium, 1 mL/min, constant
sample TBDMS derivatives of amino acids, each approximately 23 ug/mL
injection 0.5 µL, splitless (1.0 min)
liner 2 mm I.D., straight
inj. temp. 250 °C
MSD interface 325 °C
scan range m/z = 40-450
Description
Categories Analytical Chromatography, GC Applications, Amino Acids
Featured Industry Food and Beverages
Life Science and Biopharma
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Glycine
  • L-4-Hydroxyproline
  • L-Alanine
  • L-Asparagine
  • L-Aspartic acid
  • L-Cysteine
  • L-Cystine
  • L-Glutamic acid
  • L-Glutamine
  • L-Histidine
  • L-Isoleucine
  • L-Leucine
  • L-Lysine
  • L-Methionine
  • L-Phenylalanine
  • L-Proline
  • L-Serine
  • L-Threonine
  • L-Tryptophan
  • L-Tyrosine
  • L-Valine