GC Analysis of Aromatics and Alcohols on SLB®-ILD3606, Temperature Programmed Analysis
GC Analysis of Aromatics and Alcohols on SLB®-ILD3606, Temperature Programmed Analysis
Conditions
column SLB-ILD3606, 60 m x 0.25 mm I.D., 0.20 µm (29691-U)
column 250 °C
oven 50 °C (6 min), 15 °C/min to 265 °C (10 min)
sample 8 analytes at various concentrations in isooctane
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
detector FID, 250 °C
Description
Analysis Note The measurement of aromatic and oxygenate compounds in petroleum samples is a common application performed worldwide, both in on-site labs at industrial facilities and also in third-party testing labs. The SLB-ILD3606 column is able to resolve benzene (and other aromatics) from oxygenates, such as alcohols.
Categories Analytical Chromatography, GC Applications, Aromatics, Alcohols, Ketones, Hydrocarbons
Featured Industry Petroleum