GC Analysis of Background Levels of a Carbopack™ X Thermal Desorption Tube on SPB®-1
GC Analysis of Background Levels of a Carbopack™ X Thermal Desorption Tube on SPB®-1
Conditions
column SPB-1, 60 m x 0.25 mm I.D., 1.0 µm (24031)
oven 35 °C (8 min), 5 °C/min to 100 °C, 15 °C/min to 230 °C (hold 4 min)
carrier gas nitrogen, 2 mL/min. constant
injection split 10:1
inj. temp. 200 °C
MSD interface 230 °C
scan range m/z = 35-260
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Industrial Hygiene
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, Carbopack is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC