GC Analysis of Background Levels of a Tenax® TA Thermal Desorption Tube on SPB®-1
GC Analysis of Background Levels of a Tenax® TA Thermal Desorption Tube on SPB®-1
Conditions
column SPB-1, 60 m x 0.25 mm I.D., 1.0 µm (24031)
oven 35 °C (8 min), 5 °C/min to 100 °C, 15 °C/min to 230 °C (hold 4 min)
carrier gas nitrogen, 2 mL/min. constant
injection split 10:1
inj. temp. 200 °C
MSD interface 230 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Industrial Hygiene
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, Tenax is a registered trademark of Buchem B.V.
Other Notes Complementary products:
Syringes