GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Conditions
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven 150 °C (4 min), 4 °C/min to 250C (5 min)
carrier gas helium, 20 cm/sec @ 150 °C
sample 100 ng on-column of a 26 component BAME standard (47080-U)
injection 1 µL, split 100:1
liner split, cup design
inj. temp. 250 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Bacterial Acid Methyl Esters (BAMEs)
Featured Industry Clinical
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Methyl 12-methyltetradecanoate
  • Methyl 13-methyltetradecanoate
  • Methyl 14-methylpentadecanoate
  • Methyl 15-methylhexadecanoate
  • Methyl 2-hydroxydecanoate
  • Methyl 2-hydroxydodecanoate
  • Methyl 2-hydroxyhexadecanoate
  • Methyl 2-hydroxytetradecanoate
  • Methyl 3-hydroxydodecanoate
  • Methyl 3-hydroxytetradecanoate
  • Methyl cis-9-hexadecenoate
  • Methyl dodecanoate
  • Methyl eicosenoate
  • Methyl heptadecanoate
  • Methyl hexadecenoate
  • Methyl nonadecanoate
  • Methyl pentadecanoate
  • Methyl tetradecanoate
  • Methyl tridecanoate
  • Methyl undecanoate
  • Methyl cis-9,10-methylenehexadecanoate
  • Methyl cis-9,10-methyleneoctadecanoate
  • Methyl cis-9,12-octadecadienoate
  • Methyl cis-9-octadecanoate
  • Methyl octadecanoate
  • Methyl trans-9-octadecenoate