GC Analysis of Barbiturate Enantiomers on β-DEX™ 120
GC Analysis of Barbiturate Enantiomers on β-DEX™ 120
Conditions
column beta-DEX 120, 30 m x 0.25 mm I.D., 0.25 µm (24304)
oven 210 °C
carrier gas helium, 20 cm/sec @ 275 °C
injection 100:1 split
inj. temp. 300 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Chiral Applications, Clinical, Forensic & Toxicology, Enantiomers, Drugs of Abuse
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)
Legal Information DEX is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Hexobarbital
  • Mephobarbital