Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Basic Drugs on SLB®-5ms
GC Analysis of Basic Drugs on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.50 µm (28473-U)
oven 45 C (2.0 min), 30 C/min to 110 C (1.0 min), 15 C/min to 200 C (1.0 min), 4 C/min to 310 C (5.0 min)
carrier gas helium, 0.7 ml/min constant
sample drugs of abuse standard diluted to 100 ppm in methanol
injection 0.5 µL, pulsed splitless, 30 psi (0.20 min), purge on (1.50 min), purge flow (50.0 ml/min)
liner 2 mm I.D., straight
inj. temp. 250 C
MSD interface 330 C
scan range m/z = 40-450
Description
Categories Analytical Chromatography, GC Applications, Drugs of Abuse, Pharmaceuticals
Featured Industry Forensics and Toxicology
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Amitriptyline
  • Caffeine
  • Cocaine
  • Codeine
  • Desipramine
  • Diazepam
  • Diphenhydramine
  • Fentanyl
  • Heroin
  • Lidocaine
  • Methamphetamine
  • Methodone
  • Morphine
  • Nicotine
  • Phenobarbital
  • Prazepam