GC Analysis of Bitter Orange Essential Oil on SLB®-5ms
GC Analysis of Bitter Orange Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50-250C (10 min) at 3C/min
carrier gas He at 99.5 kPa (30.0 cm/s)
sample cold-pressed oils obtained from the peel of the fruits
injection split; split ratio,1:100; injected volume, 1.0 µL of diluted oil
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes