GC Analysis of α-Bromo-γ-Butyrolactone Enantiomers on Astec® CHIRALDEX® G-DP
GC Analysis of α-Bromo-γ-Butyrolactone Enantiomers on Astec® CHIRALDEX® G-DP
Conditions
column Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 µm (78033AST)
oven 160 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: alpha-bromo-gamma-butyrolactone enantiomers
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Esters, Enantiomers, Lactones
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes