GC Analysis of C1-C5 Hydrocarbons on Alumina Sulfate PLOT
GC Analysis of C1-C5 Hydrocarbons on Alumina Sulfate PLOT
Conditions
column Alumina sulfate PLOT, 50 m x 0.53 mm I.D. (28324-U)
column temp. 200 °C
oven 35 °C (2.5 min), 5 °C/min to 150 °C (4.5 min)
carrier gas helium, 3.0 mL/min
sample 18-component C1-C5 hydrocarbon standard in nitrogen, each analyte at 25 ng on-column
injection 10 µLdirect using a sampling valve, 150 °C
detector FID, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Light Hydrocarbons, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
suitability application for GC
Analyte
  • 1,3-Butadiene
  • 1-Butene
  • Acetylene
  • cis-2-Butene
  • Cyclopropane
  • Ethane
  • Ethylene
  • Isobutane
  • Isobutylene
  • Isopentane
  • Methane
  • n-Butane
  • n-Pentane
  • Propadiene
  • Propane
  • Propylene
  • Propyne
  • trans-2-Butene