GC Analysis of Camphor Enantiomers on Astec® CHIRALDEX® G-DP
GC Analysis of Camphor Enantiomers on Astec® CHIRALDEX® G-DP
Conditions
column Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 µm (78033AST)
oven 100 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: camphor enantiomers
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Volatiles
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Food and Beverages
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes