GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SLB®-IL100
GC Analysis of cis/trans C18:1, C18:2, and C18:3 FAME Isomers on SLB®-IL100
Conditions
column SLB-IL100, 60 m x 0.25 mm I.D., 0.20 µm (28886-U)
oven 170 °C
carrier gas helium, 30 cm/sec
sample Mixture of C18:1, C18:2, and C18:3 FAMEs in methylene chloride
injection 1 µL50:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes