GC Analysis of cis/trans Limonene Oxide Enantiomers on Astec® CHIRALDEX® B-PH
GC Analysis of cis/trans Limonene Oxide Enantiomers on Astec® CHIRALDEX® B-PH
Conditions
column Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 µm (71023AST)
oven 90 °C
carrier gas helium, 24 psi
sample 3 mg/mL in methylene chloridepeak 1: d-cis or d-trans limonene oxidepeak 2: l-cis and trans limonene oxidepeak 3: d-trans or d-cis limonene oxide
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Terpenes, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes