GC Analysis of Class 2 Residual Solvents on SUPELCOWAX® 10
GC Analysis of Class 2 Residual Solvents on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30 m x 0.53 mm I.D., 1.0 µm (25301-U)
oven 35 °C (15 min), 5 °C/min to 200 °C
carrier gas helium, 30 cm/sec @ 35 °C
sample 28 analytes, at concentrations indicated in DMSO
injection 1 µL, 33:1 split
liner single taper
inj. temp. 225 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Pharmaceutical (small molecule)
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC