GC Analysis of Class 2 Residual Solvents on OVI-G43
GC Analysis of Class 2 Residual Solvents on OVI-G43
Conditions
column OVI-G43, 30 m x 0.53 mm I.D., 3.0 µm (25396)
oven 35 °C (15 min), 5 °C/min to 200 °C
carrier gas helium, 30 cm/sec @ 35 °C
sample 28 analytes, at concentrations indicated in DMSO
injection 1 µL, 33:1 split
liner single taper
inj. temp. 225 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Pharmaceutical (small molecule)
suitability application for GC
Analyte
  • 1,2-Dimethoxyethane
  • 1,4-Dioxane
  • 2-Ethoxyethanol
  • 2-Methoxyethanol
  • Acetonitrile
  • Chlorobenzene
  • Chloroform
  • cis-1,2-Dichloroethylene
  • Cyclohexane
  • Dimethylacetamide
  • Dimethylformamide (DMF)
  • Ethylene glycol
  • Formamide
  • Hexane
  • Methanol
  • Methyl butyl ketone
  • Methylcyclohexane
  • Methylene chloride
  • m-Xylene
  • Nitromethane
  • N-Methylpyrrolidone
  • o-Xylene
  • p-Xylene
  • Pyridine
  • Sulfolane
  • Tetralin
  • Toluene
  • Trichloroethylene