GC Analysis of Cold Pressed Lemon Essential Oil on Equity®-1 and Equity®-5
GC Analysis of Cold Pressed Lemon Essential Oil on Equity®-1 and Equity®-5
Conditions
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
column temp. 75 °C (8 min) to 200 °C @ 4 °C/min (10 min)
flow rate helium, 30 cm/sec @ 110 °C
sample cold pressed lemon oil
injection wet needle, split 100:1
liner split, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 3-Carene
  • 6-Methyl-5-hepten-2-one
  • Camphene
  • Carvone
  • Citronellal
  • Citronellyl acetate
  • Decanal
  • Dodecanal
  • Geranial
  • Geraniol
  • Geranyl acetate
  • Heptanal
  • Linalool
  • Myrcene
  • Neral
  • Nerol
  • Neryl acetate
  • Nonanal
  • Nonyl acetate
  • Octanol
  • Octanol
  • Octyl acetate
  • o-Limonene
  • p-Cymene
  • Sabinene
  • Terpinen-4-ol
  • Terpinolene
  • trans-α-Bergansotene
  • α-Humulene
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Thujene
  • β-Bisabolene
  • β-Caryophyllene
  • β-Pinene
  • γ-Terpinene