GC Analysis of Conjugated Linoleic Acid (CLA) FAME Isomers on SP™-2560
GC Analysis of Conjugated Linoleic Acid (CLA) FAME Isomers on SP™-2560
Conditions
column SP-2560, 200 meters x 0.25mm ID x 0.2um (29688-U)
oven 175 °C
carrier gas hydrogen
sample 4-component CLA FAME isomer mix, 0.5 mg/mL in methylene chloride
injection 1µL, split 50:1
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Four key conjugated linoleic acid (CLA) FAME isomers are resolved.
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC