GC Analysis of Deprenyl (Selegiline) Enantiomers on Astec® CHIRALDEX® B-DM
GC Analysis of Deprenyl (Selegiline) Enantiomers on Astec® CHIRALDEX® B-DM
Conditions
column Astec CHIRALDEX B-DM, 30 m x 0.25 mm I.D., 0.12 µm (77023AST)
oven 130 °C
carrier gas helium, 24 psi
sample peak 1: R(-)-deprenyl (Selegiline)peak 2: S(+)-deprenyl
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Chiral Applications, Pharmaceutical (small molecule), Clinical, Forensic & Toxicology, Enantiomers, Pharmaceuticals, Therapeutic Drug Monitoring, Antiparkinson Drugs
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes