GC Analysis of Distilled Lime Essential Oil on Equity®-1 and Equity®-5
GC Analysis of Distilled Lime Essential Oil on Equity®-1 and Equity®-5
Conditions
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
column Equity-1, 30 m x 0.25 mm I.D., 0.25 µm (28046-U)
oven 75 °C (8 min), 4 °C/min to 200 °C (10 min)
carrier gas helium, 30 cm/sec @ 110 °C
sample distilled lime oil
injection Wet Needle, 100:1 split
liner split, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,4-Cineole
  • Borneol
  • Camphene
  • Decanal
  • d-Limonene
  • Dodecanal
  • Geranial
  • Geranyl acetate
  • Linalool
  • Myrcene
  • Neral
  • Neryl acetate
  • p-Cymene
  • Terpinen-1-ol
  • Terpinen-4-ol
  • Terpinolene
  • trans-α-Bergamotene
  • trans-α-Farensene
  • α-Fenchyl alcohol
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • β-Bisabolene
  • β-Caryophyllene
  • β-Pinene
  • β-Terpineol
  • γ-Terpinene
  • γ-Terpineol