GC Analysis of FAMEs in Rapeseed Oil (with Conjugated Linoleic Acid (CLA) FAME Isomers Added) on SLB®-IL111
GC Analysis of FAMEs in Rapeseed Oil (with Conjugated Linoleic Acid (CLA) FAME Isomers Added) on SLB®-IL111
Conditions
column SLB-IL111, 200 m x 0.25 mm I.D., 0.20 µm (29689-U)
oven 150 °C
carrier gas hydrogen
sample Mix of rapeseed oil FAME isomers and CLA FAME isomers
injection 1µL, split 50:1
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Four key conjugated linoleic acid (CLA) FAME isomers are resolved in a mixture with rapeseed oil FAME isomers. The SLB-IL111 provides a different elution pattern compared to SP-2560 (a cyanopropyl siloxane column).
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture