GC Analysis of Fluorocarbons on Alumina Chloride PLOT
GC Analysis of Fluorocarbons on Alumina Chloride PLOT
Conditions
column Alumina Chloride PLOT, 50m x 0.53mm ID, 10mum (28329-U)
oven 35 °C (1.0 min), 12 °C/min to 180 °C
carrier gas helium, 3.0 mL/min
sample Freons @ ~20% each (custom standard)
injection 10 µL
inj. temp. 200 °C
detector FID, 230C
Description
Categories Analytical Chromatography, GC Applications, Gases
Featured Industry Chemicals and Industrial Polymers
suitability application for GC
Analyte
  • Dichlorofluoromethane
  • Freon 11
  • Freon 114
  • Freon 114a
  • Freon 12
  • Freon 21
  • Freon 22
  • Trichlorofluoromethane