GC Analysis of Fragrance Compounds on SUPELCOWAX® 10
GC Analysis of Fragrance Compounds on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µm (24079)
oven 50 °C (2 min), 2 °C/min to 280 °C (20 min)
carrier gas helium, 25 cm/sec @ 200 °C
inj. temp. 0.2 µLsplit (100:1)
detector FID
Description
Categories Analytical Chromatography, GC Applications, Volatiles
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (-)-Bornyl acetate
  • (-)-Menthone
  • 1,8-Cineole
  • 6-Methyl-5-hepten-2-one
  • Benzyl acetate
  • Camphene
  • Cedrol, redistilled
  • Cedryl acetate
  • Cinnamyl cinnamate
  • cis-3,7-Dimethyl-2,6-octadien-1-ol
  • cis-Jasmone
  • Coumarin
  • Decanal
  • Ethanol
  • Ethyl acetate
  • Ethyl behenate
  • Ethyl stearate
  • Geranyl acetate
  • Limonene
  • Linalool
  • Linalyl acetate
  • Linalyl acetate
  • Methyl jasmonate
  • Musk ambrette
  • Musk ketone
  • Musk xylol
  • Myrcene
  • Nerolidol
  • Nonanal
  • Octanal
  • Phytol
  • Vanillin
  • α-Ionone
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Thujene
  • β-Caryophyllene
  • β-Pinene