GC Analysis of Fuel Oil #6 on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Fuel Oil #6 on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 80 °C, 50 °C/min to 325 °C
carrier gas hydrogen, 45 cm/sec constant
sample No.6 Fuel Oil standard, 20 mg/mL in hexane:chloroform (47536-U)
injection 0.3 µL, 100:1 split, 0.1 min pre-injection dwell time
liner 2 mm I.D., straight
inj. temp. 250 °C
detector FID, 350 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes