GC Analysis of Glycidol Enantiomers on Astec® CHIRALDEX® B-TA
GC Analysis of Glycidol Enantiomers on Astec® CHIRALDEX® B-TA
Conditions
column Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 µm (73023AST)
oven 80 °C
carrier gas helium, 30 psi
sample peak 1: S(-)-glycidol peak 2: R(+)-glycidol
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Elution order reversal on CHIRALDEX A-TA and CHIRALDEX G-TA.
Categories Analytical Chromatography, GC Applications, Alcohols, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes