GC Analysis of Impurities in Propylene on Carboxen®-1006 PLOT
GC Analysis of Impurities in Propylene on Carboxen®-1006 PLOT
Conditions
column Carboxen-1006 PLOT, 30 m x 0.32 mm I.D. (24241-U)
oven 200 °C (4 min), 24 °C/min to 250 °C
carrier gas helium, 76 cm/sec
sample 6 impurities in propylene, each at approximately 1.0%
injection 20 µL
inj. temp. 230 °C
detector FID, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Light Hydrocarbons, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acetylene
  • Ethane
  • Ethylene
  • Methane
  • Propane
  • Propylene
  • Propyne