GC Analysis of Industrial Solvents on Equity®-1701
GC Analysis of Industrial Solvents on Equity®-1701
Conditions
column Equity-1701, 30 m x 0.32 mm I.D., 1.0 µm (28199-U)
oven 35 °C (8 min), 4 °C/min to 130 °C (2 min)
carrier gas helium, 25 cm/sec
sample 59-Component neat solvent mix
injection 0.5 µL200:1 split
liner Split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethylene
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,4-Dioxane
  • 1-Propanol
  • 2-Butanone
  • 2-Ethoxyethanol
  • 2-Ethoxyethyl acetate
  • 2-Hexanone
  • 2-Methoxyethanol
  • 2-Methylcyclohexanol isomers
  • 2-Methylphenol
  • 2-Nitropropane
  • 2-Propanol
  • 3-Methylphenol
  • 4-Methyl-2-pentanone
  • 4-Methylphenol
  • 5-Methyl-2-hexanone
  • Acetone
  • Benzene
  • Butyl Cellosolve
  • Carbon tetrachloride
  • Chlorobenzene
  • Chloroform
  • Cyclohexanol
  • Diacetone alcohol
  • Dimethyl formamide
  • Ethanol
  • Ethyl acetate
  • Ethyl formate
  • Ethylbenzene
  • Hexane
  • Isoamyl acetate
  • Isoamyl alcohol
  • Isobutanol
  • Isobutyl acetate
  • Isopropyl acetate
  • Mesityl oxide
  • Methanol
  • Methyl acetate
  • Methyl formate
  • Methylene chloride
  • m-Xylene
  • n-Amyl acetate isomers
  • n-Butanol
  • n-Butyl acetate
  • n-Propyl acetate
  • o-Xylene
  • p-Xylene
  • sec-Butanol
  • Styrene
  • Tetrachloroethene
  • Tetrahydrofuran (THF)
  • Toluene
  • trans-1,2-Dichloroethylene
  • Trichloroethylene