GC Analysis of Ionic Liquid Test Mix on SLB®-IL100 (15 m x 0.10 mm I.D., 0.08 μm)
GC Analysis of Ionic Liquid Test Mix on SLB®-IL100 (15 m x 0.10 mm I.D., 0.08 μm)
Conditions
column SLB-IL100, 15 m x 0.10 mm I.D., 0.08 µm (28882-U)
oven 90 °C
carrier gas hydrogen, 50 cm/sec
sample Ionic Liquid Test Mix #1, each analyte at various concentrations in isooctane (N09000)
inj. temp. 250 °C
detector FID, 250 °C
detector 0.5 µL300:1 split
Description
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • Cyclohexanone
  • Ethylbenzene
  • Isooctane
  • Isopropylbenzene (cumene)
  • Naphthalene
  • n-Tridecane
  • p-Xylene
  • Toluene