GC Analysis of Lavender Spike Essential Oil on SLB®-5ms
GC Analysis of Lavender Spike Essential Oil on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 °C, 3 °C/min to 280 °C (5 min)
carrier gas helium, 30 cm/s
sample Lavender spike essential oil
injection 1 µL200:1 split
inj. temp. 280 °C
detector MSD
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances