GC Analysis of Lavender Spike Essential Oil on SLB®-IL60
GC Analysis of Lavender Spike Essential Oil on SLB®-IL60
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 50 °C, 3 °C/min to 270 °C (5 min)
carrier gas helium, 30 cm/s
sample Lavender spike essential oil
injection 0.5 µL50:1 split
inj. temp. 270 °C
detector FID, 270 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances