GC Analysis of Nitrosamines in Water on PTA-5 after SPME using 65 μm PDMS/DVB Fiber
GC Analysis of Nitrosamines in Water on PTA-5 after SPME using 65 μm PDMS/DVB Fiber
Conditions
sample/matrix analytes in water +25% KCl, pH 10.0
desorption process 270 °C for 1 min
extraction immersion, 15 min (rapid stirring)
SPME fiber 65 μm polydimethylsiloxane/divinylbenzene (57310-U)
column PTA-5, 30 m x 0.32 mm I.D., 0.5 µm (24331)
oven 50 °C (1 min), 10 °C/min to 250 °C (2 min)
carrier gas helium, 30 cm/sec
liner 0.75 mm I.D., SPME
inj. temp. 250 °C
detector MSD, SIM
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Amines & Nitroaromatics, Amines
Featured Industry Chemicals and Industrial Polymers
Environmental
Food and Beverages
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC
Analyte
  • Nitrosodibutylamine
  • Nitrosodiethylamine
  • Nitrosodimethylamine
  • Nitrosodipropylamine
  • Nitrosomethylethylamine
  • Nitrosopiperidine
  • N-Nitrosodiphenylamine