GC Analysis of PCB Congeners in Stream Sediment on SPB®-Octyl after SPME using 100 μm PDMS Fiber
GC Analysis of PCB Congeners in Stream Sediment on SPB®-Octyl after SPME using 100 μm PDMS Fiber
Conditions
sample/matrix 5 g stream sediment, downstream from industrial sites
desorption process 280 °C/2 min, splitless
extraction headspace, 90 °C/30 min
SPME fiber PDMS, 100 µm (57300-U) (57300-U)
column SPB®-Octyl, 30 m x 0.25 mm I.D., 0.25 µm (24218-U)
oven 50 °C/1 min at 10 °C/min to 150 °C/0 min
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Environmental, Dioxins, PCBs, PBDEs, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC