GC Analysis of PCBs in Transformer Oil on a 5%Phenyl Column after SPE using Supelclean™ Sulfoxide
GC Analysis of PCBs in Transformer Oil on a 5%Phenyl Column after SPE using Supelclean™ Sulfoxide
Conditions
sample preparation SPE (Solid Phase Extraction)
extraction process Sample Pre-treatment: paraffin oil was spiked with a Kanechlor PCB mix at the total levels of 3.7 ppm (mg/kg) and diluted with hexane (1:1 v/v). (Commercial insulation oil (Japan Industrial Standard JIS C2320-1999, insulating oil, Class 1-2/4).)
SPE tube/cartridge Supelclean Sulfoxide SPE Tube, Glass 6 g/20 mL (55252-U)
condition 20 mL acetone; 40 mL of hexane
sample addition transformer oil spiked with PCBs
sample addition 0.4 mL diluted oil sample
elution 12 mL hexane (elutes oil interferences); 25 mL hexane (elutes PCBs)
eluate post-treatment concentrate under nitrogen
column 5% phenyl polysiloxane (Supelco equivalent, SLB-5ms, 28471-U)
detector QMS (For full method details, please see reference (1))
Description
Application Sample Pre-treatment:
Commercial insulation oil (Japan Industrial Standard JIS C2320-1999, insulating oil, Class 1-2/4, paraffin oil) was spiked with a Kanechlor PCB mix at the total levels of 3.7 ppm (mg/kg) and diluted with hexane (1:1 v/v).
Categories Analytical Chromatography, SPE Applications, GC Applications, Dioxins, PCBs, PBDEs, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information Supelclean is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE