GC Analysis of Permanent Gases on Mol Sieve 5A PLOT
GC Analysis of Permanent Gases on Mol Sieve 5A PLOT
Conditions
column Mol Sieve 5A PLOT, 30 m x 0.53 mm I.D. (25463)
oven 65 °C
carrier gas helium, 10.0 mL/min
sample Oxygen, methane, and carbon monoxide, each at 1.0% in nitrogen
injection 10.0 mu, direct using a sample valve
inj. temp. 200 °C
detector TCD, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Gases, Volatiles