GC Analysis of Permanent Gases and Light Hydrocarbons on Carboxen®-1006 PLOT
GC Analysis of Permanent Gases and Light Hydrocarbons on Carboxen®-1006 PLOT
Conditions
column Carboxen-1006 PLOT, 30 m x 0.53 mm I.D. (25461)
oven 35 °C (1.0 min), 24 °C/min to 250 °C
carrier gas helium, 10.0 mL/min
sample Permanent gas and light hydrocarbon mix, analytes at concentrations indicated
injection 10.0 µL, direct using a sample valve, 150 °C
inj. temp. 200 °C
detector TCD, 230 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Light Hydrocarbons, Hydrocarbons, Gases
Featured Industry Chemicals and Industrial Polymers
Legal Information Carboxen is a registered trademark of Sigma-Aldrich Co. LLC