GC Analysis of Permanent Gases and Light Hydrocarbons on Supel-Q™ PLOT
GC Analysis of Permanent Gases and Light Hydrocarbons on Supel-Q™ PLOT
Conditions
column Supel-Q PLOT, 30 m x 0.53 mm I.D. (25462)
oven 35 °C (3 min), 16 C/min to 250 °C
carrier gas helium, 3 mL/min
sample 9-Component permanent gas a light hydrocarbon standard, each analyte at 1.0% in nitrogen
injection 0.1 µL
inj. temp. 200 °C
detector TCD, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Volatiles, Light Hydrocarbons, Gases, Hydrocarbons
Featured Industry Chemicals and Industrial Polymers
Legal Information Supel-Q is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acetylene
  • Carbon dioxide
  • Ethane
  • Ethylene
  • Methane
  • n-Butane
  • Nitrogen
  • Propane
  • Propylene
  • Propyne
  • Water