GC Analysis of Pesticides in Grapes on SLB®-5ms after QuEChERS Cleanup using Supel™ QuE PSA/C18
GC Analysis of Pesticides in Grapes on SLB®-5ms after QuEChERS Cleanup using Supel™ QuE PSA/C18
Conditions
sample preparation Dispersive (QuEChERS)
sample/matrix Thompson green seedless table grapes
extraction tube Supel QuE Acetate extraction tube (55234-U)
extraction process homogenize, add 15 mL of acetonitrile with 1% acetic acid; shake for 1 minute; add contents of Supel QuE acetate extraction tube; shake at 2500 rpm for 10 minutes
clean-up process transfer 1 mL of acetonitrile layer into cleanup tube; shake for 1 minute; centrifuge at 5000 rpm for 3 minutes; draw off supernatant for LVI-GC/MS analysis
clean-up tube Supel QuE PSA/C18 suitable for 2007.01 per AOAC (55288-U)
column SLB-5ms, 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
oven 50 °C (2.9 min), 8 °C/min to 330 °C
carrier gas helium, 0.7 mL/min constant flow
injection 10 µL PTV; 100 mL/min vent flow (5 psi) until 0.4 min, purge flow to split vent 60 mL/min at 2.9 min
liner 4 mm ID Focus Liner w/taper
inj. temp. programmed; 50 °C (0.4 min), 600 °C/min to 325 °C (5 min)
detector MS-SIM
MSD interface 330 °C
scan range SIM
Description
Application The "Quick, Easy, Cheap, Effective, Rugged, and Safe" or QuEChERS approach was used to extract pesticides incurred in Thompson green seedless grapes. The presence of several pesticides commonly used on grapes was identified.
Categories Analytical Chromatography, SPE Applications, GC Applications, Pesticides
Featured Industry Agriculture
Food and Beverages
Legal Information Supel is a trademark of Sigma-Aldrich Co. LLC, SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Sample pretreatment: homogenize
suitability application for GC, application for SPE
Analyte
  • Cyprodinil
  • Myclobutanil
  • Quinoxyfen
  • Tebuconazole